missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dichloronicotinic acid, 97%, Thermo Scientific™

Product Code. 15204784 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
Unit Size:
1g
This item is not returnable. View return policy

Product Code. 15204784

Brand: Thermo Scientific Acros 449830010

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 262423-77-8
Melting Point 151°C to 156°C
Assay Percent Range 97%
Packaging Glass bottle
Molecular Formula C6H3Cl2NO2
MDL Number MFCD06796292
Quantity 1 g
Synonym 2,4-dichloronicotinic acid, 2,4-dichloronicotinc acid, 2,4-dichloronicotinicacid, 2,4-dichloro-3-pyridinecarboxylic acid, 2.4-dichloronicotinic acid, 2,4-dichloropyridine-3-carboxylicacid, dichloronicotinic acid;, pubchem17746, 2,4-dichloronicotinic acid;
InChI Key ZFGZNVSNOJPGDV-UHFFFAOYSA-N
SMILES OC(=O)C1=C(Cl)C=CN=C1Cl
IUPAC Name 2,4-dichloropyridine-3-carboxylic acid
Molecular Weight (g/mol) 192.00
PubChem CID 10583887
Formula Weight 192
Percent Purity 97%
Chemical Name or Material 2, 4-Dichloronicotinic acid
Show More Show Less
compliance-icons
Product Identifier
  • 2,4-Dichloronicotinic acid
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.