Learn More
2,4-Dichloronicotinic acid, 97%, Thermo Scientific™
Product Code. 15494558
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
Product Code. 15494558
Brand: Thermo Scientific Alfa Aesar H58239.MD
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 262423-77-8
Molecular Formula C6H3Cl2NO2
Molecular Weight (g/mol) 192.00
MDL Number MFCD06796292
InChI Key ZFGZNVSNOJPGDV-UHFFFAOYSA-N
Synonym 2,4-dichloronicotinic acid, 2,4-dichloronicotinc acid, 2,4-dichloronicotinicacid, 2,4-dichloro-3-pyridinecarboxylic acid, 2.4-dichloronicotinic acid, 2,4-dichloropyridine-3-carboxylicacid, dichloronicotinic acid;, pubchem17746, 2,4-dichloronicotinic acid;
PubChem CID 10583887
IUPAC Name 2,4-dichloropyridine-3-carboxylic acid
SMILES OC(=O)C1=C(Cl)C=CN=C1Cl
Specifications
Melting Point 166°C to 167°C
Quantity 250 mg
Formula Weight 192
Percent Purity 97%
Chemical Name or Material 2,4-Dichloronicotinic acid