Learn More
2,4-Dichlorobenzonitrile, Sigma-Aldrich™
Specifications
Specifications
| CAS | 6574-98-7 |
| Melting Point | 59°C to 62°C |
| MDL Number | MFCD00016373 |
| Quantity | 100 g |
| Synonym | 2,4-Dichlorobenzonitrile, 2,4-Dichlorobenzonitrile |
| Molecular Weight (g/mol) | 172.01 |
| Grade | Research |