Learn More
2,4-Dichloro-6-methylbenzylamine, 98+%, Thermo Scientific™
Product Code. 11322897
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11322897 1 g 1g
11342897 250 mg 250mg
11332897 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11322897 Supplier Thermo Scientific Alfa Aesar Supplier No. L16108.03
 Restricted Item
Estimated Shipment: 28-07-2026
Add to basket
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 150517-76-3
Molecular Formula C8H9Cl2N
Molecular Weight (g/mol) 190.07
MDL Number MFCD00052913
InChI Key QISTWSMZJRTDCS-UHFFFAOYSA-N
Synonym 2,4-dichloro-6-methylbenzylamine, 2,4-dichloro-6-methylphenyl methanamine, 2,4-dichloro-6-methylphenyl methylamine, benzenemethanamine,2,4-dichloro-6-methyl, benzenemethanamine, 2,4-dichloro-6-methyl, 2,4-dichloro-6-methylbenzyl amine hydrochloride, 4,6-dichloro-2-methylbenzylamine, acmc-1c3je, 2-aminomethyl-3,5-dichlorotoluene, 1-2,4-dichloro-6-methylphenyl methanamine
PubChem CID 2800971
SMILES CC1=CC(Cl)=CC(Cl)=C1CN
Specifications
Melting Point 52°C to 56°C
Quantity 1 g
UN Number UN3259
Beilstein 8615227
Sensitivity Air Sensitive
IUPAC Name (2,4-dichloro-6-methylphenyl)methanamine
Formula Weight 190.07
Percent Purity ≥98%
Chemical Name or Material 2,4-Dichloro-6-methylbenzylamine