missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dibromotoluene, 98%, Thermo Scientific™

Product Code. p-7055323
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15460809 1 g 1g
15470809 5 g 5g
15480809 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15460809 Supplier Thermo Scientific Alfa Aesar Supplier No. H63869.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 31543-75-6
Molecular Formula C7H6Br2
Molecular Weight (g/mol) 249.933
MDL Number MFCD00052985
InChI Key GHWYNNFPUGEYEM-UHFFFAOYSA-N
Synonym 2,4-dibromotoluene, benzene, 2,4-dibromo-1-methyl, 2,4-bromotoluene, 2,4-dibromo-toluene, pubchem3915, 2,4-dibromo toluene, acmc-1bn9b, ksc497e7f, 2,4-dibromo-1-methyl-benzene, ghwynnfpugeyem-uhfffaoysa
PubChem CID 94421
IUPAC Name 2,4-dibromo-1-methylbenzene
SMILES CC1=C(C=C(C=C1)Br)Br

Specifications

Melting Point -10°C (decomposition)
Density 1.85 g/mL
Boiling Point 243°C (decomposition)
Refractive Index 1.601
Quantity 1 g
Beilstein 2078741
Formula Weight 249.93
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 2,4-Dibromotoluene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.