missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dibromobenzyl alcohol, 98%, Thermo Scientific™

Product Code. 15432059
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15432059 250 mg 250mg
15412059 1 g 1g
15422059 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15432059 Supplier Thermo Scientific Alfa Aesar Supplier No. H64741.MD
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
It is an halogenated compound for proteomics research. Also used as a intermediate for pharmaceutical.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 666747-06-4
Molecular Formula C7H6Br2O
Molecular Weight (g/mol) 265.93
MDL Number MFCD09996906
InChI Key VXEVXBMLHQPKGA-UHFFFAOYSA-N
PubChem CID 17836195
IUPAC Name (2,4-dibromophenyl)methanol
SMILES OCC1=C(Br)C=C(Br)C=C1

Specifications

Quantity 250 mg
Beilstein 3241294
Synonym 2,4-dibromophenyl methanol, 2,4-dibromobenzyl alcohol, benzenemethanol, 2,4-dibromo, 2,4-dibromobenzenemethanol, benzenemethanol,2,4-dibromo, pubchem9781, 2,4-dibromobenzylalcohol, acmc-1bfu5, 2,4-bis bromanyl phenyl methanol
Solubility Information Slightly soluble in water.
Formula Weight 265.98
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 2,4-Dibromobenzyl alcohol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.