Learn More
2,4-Diamino-6-piperidinopyrimidine (Desoxyminoxidil), TRC
Product Code. 30003627
Change view
Click to view available options
Quantity:
100 mg
250 mg
500 mg
Unit Size:
100mg
250mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30003627 500 mg 500mg
30007673 250 mg 250mg
17982585 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30003627 Supplier Toronto Research Chemicals Supplier No. TRCD416640500MG
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 24867-26-3
Molecular Formula C9 H15 N5
Molecular Weight (g/mol) 193.25
Synonym 6-(Piperidin-1-yl)pyrimidine-2,4-diamine, Minoxidil Imp. E (EP), Desoxyminoxidil
IUPAC Name 6-piperidin-1-ylpyrimidine-2,4-diamine
SMILES Nc1cc(nc(N)n1)N2CCCCC2
Specifications
Quantity 500 mg
InChI Formula InChI=1S/C9H15N5/c10-7-6-8(13-9(11)12-7)14-4-2-1-3-5-14/h6H,1-5H2,(H4,10,11,12,13)
Formula Weight 193.1327
Percent Purity >95%
Chemical Name or Material 6-(Piperidin-1-yl)pyrimidine-2,4-diamine (Desoxyminoxidil)
Grade HPLC