Learn More
2-(4-Chlorobenzoyl)pyridine, Mikromol™
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
Description
2-(4-Chlorobenzoyl)pyridine
Specifications
Specifications
| Chemical Name or Material | 2-(4-Chlorobenzoyl)pyridine |
| CAS | 6318-51-0 |
| Analyte or Component Names | 2-(4-Chlorobenzoyl)pyridine |
| Shipping Condition | Room Temperature |
| Molecular Formula | C12 H8 Cl N O |
| Certifications/Compliance | ISO 17025 |
| Synonym | (4-Chlorophenyl)-2-pyridinylmethanone, Ketone, p-chlorophenyl 2-pyridyl (6CI,8CI), (4-Chlorophenyl)-2-pyridinylmethanone, 2-(4-Chlorobenzoyl)pyridine, 2-(p-Chlorobenzoyl)pyridine, NSC 31646, Carbinoxamine Related Compound A (USP), Carbinoxamine Related Compound A |
| InChI Formula | InChI=1S/C12H8ClNO/c13-10-6-4-9(5-7-10)12(15)11-3-1-2-8-14-11/h1-8H |
| SMILES | Clc1ccc(cc1)C(=O)c2ccccn2 |
| IUPAC Name | (4-chlorophenyl)-pyridin-2-ylmethanone |
| Show More |