Learn More
2-(4-Bromophenyl)-6-iodoimidazo[1,2-a]pyridine, 97%, Thermo Scientific™
Product Code. 15458005
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15458005 5 g 5g
15448005 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15458005 Supplier Thermo Scientific Alfa Aesar Supplier No. H32156.06
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 118000-66-1
Molecular Formula C13H8BrIN2
Molecular Weight (g/mol) 399.029
MDL Number MFCD00444892
InChI Key DOUYIPIXURZQRX-UHFFFAOYSA-N
Synonym 2-4-bromophenyl-6-iodoimidazo 1,2-a pyridine, acmc-20aopu, 2-4-bromophenyl-6-iodoimidazo 1,2-pyridine, 2-4-bromophenyl-6-iodoh-imidazo 1,2-a pyridine, imidazo 1,2-a pyridine,2-4-bromophenyl-6-iodo, 2-4-bromophenyl-6-iodoimidazo 1 2-a p
PubChem CID 14042758
IUPAC Name 2-(4-bromophenyl)-6-iodoimidazo[1,2-a]pyridine
SMILES C1=CC(=CC=C1C2=CN3C=C(C=CC3=N2)I)Br
Specifications
Melting Point 212°C to 216°C
Quantity 5 g
Sensitivity Light sensitive
Formula Weight 399.03
Percent Purity 97%
Chemical Name or Material 2-(4-Bromophenyl)-6-iodoimidazo[1,2-a]pyridine