Learn More
2-[4-(Boc-amino)-1-piperidinyl]pyridine-5-boronic acid pinacol ester, 96%, Thermo Scientific™
Product Code. 15400896
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
Product Code. Quantity unitSize
15400896 1 g 1g
15410896 250 mg 250mg
2 options
Product Code. 15400896
Brand: Thermo Scientific Alfa Aesar H54203.03
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
2-[4-(Boc-amino)-1-piperidinyl]pyridine-5-boronic acid pinacol ester is used as an organic chemical synthesis intermediate.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents and heat.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1042917-44-1
Molecular Formula C21H34BN3O4
Molecular Weight (g/mol) 403.33
MDL Number MFCD13195039
InChI Key JGRVQSYVAOLRKS-UHFFFAOYSA-N
Synonym tert-butyl 1-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridin-2-yl piperidin-4-yl carbamate, 2-4-boc-amino-1-piperidinyl pyridine-5-boronic acid pinacol ester, tert-butyl n-1-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridin-2-yl piperidin-4-yl carbamate, 6-4-boc-amino piperidin-1-yl pyridine-3-boronic acid pinacol ester, 2-4-tert-butyloxycarbonylamino-1-piperidinyl pyridine-5-boronic acid pinacol ester, carbamic acid, n-1-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-2-pyridinyl-4-piperidinyl-, 1,1-dimethylethyl ester
PubChem CID 68484693
IUPAC Name tert-butyl N-[1-[5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-yl]piperidin-4-yl]carbamate
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CN=C(C=C2)N3CCC(CC3)NC(=O)OC(C)(C)C
Specifications
Melting Point 132°C to 134°C
Quantity 1 g
Formula Weight 403.32
Percent Purity 96%
Chemical Name or Material 2-[4-(Boc-amino)-1-piperidinyl]pyridine-5-boronic acid pinacol ester