missing translation for 'onlineSavingsMsg'
Learn More

2',4'-Bis(trifluoromethyl)acetophenone, 97%, Thermo Scientific™

Codice prodotto. 11386909
Cambia vista
Click to view available options
Quantity:
250 mg
1 g
Dimensione della confezione:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantity unitSize
11386909 1 g 1g
15433547 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Codice prodotto. 11386909 Fornitore Thermo Scientific Alfa Aesar N. del fornitore H26802.03
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Visualizzare prodotti alternativi

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Identificatori chimici

CAS 237069-82-8
Molecular Formula C10H6F6O
Molecular Weight (g/mol) 256.147
MDL Number MFCD00042495
InChI Key MHLYNSLYUWMONA-UHFFFAOYSA-N
Synonym 1-2,4-bis trifluoromethyl phenyl ethanone, 2',4'-bis trifluoromethyl acetophenone, 2,4-di trifluoromethyl acetophenone, 1-2,4-bis trifluoromethyl phenyl ethan-1-one, ethanone, 1-2,4-bis trifluoromethyl phenyl, 2,4-bis trifluoromethyl acetophenone, pubchem7002, 2,4-ditrifluoromethylacetophenone, 1-acetyl-2,4-bis trifluoromethyl benzene
PubChem CID 2736093
IUPAC Name 1-[2,4-bis(trifluoromethyl)phenyl]ethanone
SMILES CC(=O)C1=C(C=C(C=C1)C(F)(F)F)C(F)(F)F

Specifica

Refractive Index 1.42
Quantity 1 g
Formula Weight 256.15
Percent Purity 97%
Chemical Name or Material 2',4'-Bis(trifluoromethyl)acetophenone
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.