missing translation for 'onlineSavingsMsg'
Learn More

2,4,6-Trimethoxypyrimidine, 98%, Thermo Scientific™

Product Code. 10091254 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 10091254

Brand: Thermo Scientific Acros 426740010

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 13106-85-9
Molecular Formula C7H10N2O3
Molecular Weight (g/mol) 170.17
InChI Key RJVAFLZWVUIBOU-UHFFFAOYSA-N
Synonym acmc-209bkb, 2,4,6-trimethoxy-pyrimidine, pyrimidine,2,4,6-trimethoxy, rjvaflzwvuibou-uhfffaoysa, 2,4,6-trimethoxypyrimidine
PubChem CID 726938
IUPAC Name 2,4,6-trimethoxypyrimidine
SMILES COC1=CC(=NC(=N1)OC)OC

Specifications

Melting Point 51°C to 54°C
Boiling Point 232°C
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 170.17
Percent Purity 98%
Chemical Name or Material 2, 4, 6-Trimethoxypyrimidine
compliance-icons
Product Identifier
  • 2,4,6-Trimethoxypyrimidine
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.