Learn More
2,4,6-Trimethoxyaniline, 97%, Thermo Scientific™
Product Code. 10036232
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Glass/Safe-Cote
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10036232 250 mg Glass/Safe-Cote 250mg
10046132 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10036232 Supplier Thermo Scientific Maybridge Supplier No. BTB09100CB
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Chemical Identifiers
CAS 14227-17-9
Molecular Formula C9H13NO3
Molecular Weight (g/mol) 183.207
MDL Number MFCD00017165
InChI Key FNSAKXLEFPFZOM-UHFFFAOYSA-N
Synonym benzenamine, 2,4,6-trimethoxy, 2,4,6-tri-methoxyaniline, 2,4,6-trimethoxybenzenamine, 2,4,6-trimethoxy-phenylamine, 2,4,6-trimethoxy-phenyl-amine
PubChem CID 84271
IUPAC Name 2,4,6-trimethoxyaniline
SMILES COC1=CC(=C(C(=C1)OC)N)OC
Specifications
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Glass/Safe-Cote
Quantity 250 mg
Formula Weight 183.21
Percent Purity 97%
Chemical Name or Material 2, 4, 6-Trimethoxyaniline