missing translation for 'onlineSavingsMsg'
Learn More

2,4,6-Trichlorophenyl isothiocyanate, 98%, Thermo Scientific™

Product Code. 11338625
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11338625 1 g 1g
11348625 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11338625 Supplier Thermo Scientific Alfa Aesar Supplier No. L12118.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22134-07-2
Molecular Formula C7H2Cl3NS
Molecular Weight (g/mol) 238.51
MDL Number MFCD00041053
InChI Key DSXVZIOQROMOEF-UHFFFAOYSA-N
PubChem CID 123399
IUPAC Name 1,3,5-trichloro-2-isothiocyanatobenzene
SMILES C1=C(C=C(C(=C1Cl)N=C=S)Cl)Cl

Specifications

Melting Point 72°C to 76°C
Quantity 1 g
Odor Irritating
UN Number UN2923
Beilstein 880716
Synonym 2,4,6-trichlorophenyl isothiocyanate, benzene,1,3,5-trichloro-2-isothiocyanato, benzene, 1,3,5-trichloro-2-isothiocyanato, 2,4,6-trichlorophenylisothiocyanate, acmc-1cngp, 2,4,6-trichlorobenzenisothiocyanate, #
Formula Weight 238.52
Percent Purity 98%
Chemical Name or Material 2,4,6-Trichlorophenyl isothiocyanate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.