Learn More
2,4,6-Tribromoanisole, TRC
Product Code. 30136485
Change view
Click to view available options
Quantity:
5 g
10 g
100 g
Unit Size:
100g
10g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30136485 100 g 100g
30003704 5 g 5g
30006434 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30136485 Supplier Toronto Research Chemicals Supplier No. TRCT771850100G
 Restricted Item
Estimated Shipment: 01-10-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 607-99-8
Molecular Formula C7 H5 Br3 O
Molecular Weight (g/mol) 344.83
Synonym 2,4,6-Tribromoanisole, Benzene, 1,3,5-tribromo-2-methoxy- (9CI, ACI), 1,3,5-Tribromo-2-methoxybenzene (ACI), Anisole, 2,4,6-tribromo- (6CI, 7CI, 8CI), 1-Methoxy-2,4,6-tribromobenzene, 2,4,6-Tribromoanisole, Methyl 2,4,6-tribromophenyl ether, NSC 2218, TBA
IUPAC Name 1,3,5-tribromo-2-methoxybenzene
SMILES COc1c(Br)cc(Br)cc1Br
Specifications
Quantity 100 g
InChI Formula InChI=1S/C7H5Br3O/c1-11-7-5(9)2-4(8)3-6(7)10/h2-3H,1H3
Formula Weight 341.7891
Chemical Name or Material 2,4,6-Tribromoanisole