missing translation for 'onlineSavingsMsg'
Learn More

2,4,6-Tri(dimethylaminomethyl)phenol, MP Biomedicals

Product Code. 11499214 Shop All MP Biomedicals Products
Click to view available options
Quantity:
100 g
Unit Size:
50g
This item is not returnable. View return policy

Product Code. 11499214

Brand: MP Biomedicals 0521912650

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 90-72-2
Molecular Formula C15H27N3O
Quantity 100 g
Synonym 2,4,6-tris dimethylaminomethyl phenol, sumicure d, 2,4,6-tris dimethylamino methyl phenol, dmf 3, actiron nx 3, ancamine k 54, capcure eh 30, epilink 230, araldite dy 061
InChI Key AHDSRXYHVZECER-UHFFFAOYSA-N
SMILES CN(C)CC1=CC(=C(C(=C1)CN(C)C)O)CN(C)C
IUPAC Name 2,4,6-tris[(dimethylamino)methyl]phenol
Molecular Weight (g/mol) 265.401
PubChem CID 7026
Formula Weight 265.4
Chemical Name or Material 2, 4, 6-Tri(Dimethylaminomethyl)Phenol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.