missing translation for 'onlineSavingsMsg'
Learn More

2,4,5-Trifluorocinnamic acid, 97+%, Thermo Scientific™

Catalog Number 11400847
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
247170-17-8
C9H5F3O2
202.132
MFCD00061211
KFVQEUPEGKJFCW-OWOJBTEDSA-N
2,4,5-trifluorocinnamic acid, 3-2,4,5-trifluorophenyl acrylic acid, 2e-3-2,4,5-trifluorophenyl prop-2-enoic acid, 3-2,4,5-trifluoro-phenyl-acrylic acid, 2,4,5-trifluorocinnamicacid, pubchem7388, e-3-2,4,5-trifluorophenyl acrylicacid, e-3-2,4,5-trifluorophenyl-2-propenoic acid, e-3-2,4,5-trifluorophenyl prop-2-enoic acid, e-3-2,4,5-tris fluoranyl phenyl prop-2-enoic acid
5708777
(E)-3-(2,4,5-trifluorophenyl)prop-2-enoic acid
C1=C(C(=CC(=C1F)F)F)C=CC(=O)O
This item is not returnable. View return policy
247170-17-8
C9H5F3O2
202.132
MFCD00061211
KFVQEUPEGKJFCW-OWOJBTEDSA-N
2,4,5-trifluorocinnamic acid, 3-2,4,5-trifluorophenyl acrylic acid, 2e-3-2,4,5-trifluorophenyl prop-2-enoic acid, 3-2,4,5-trifluoro-phenyl-acrylic acid, 2,4,5-trifluorocinnamicacid, pubchem7388, e-3-2,4,5-trifluorophenyl acrylicacid, e-3-2,4,5-trifluorophenyl-2-propenoic acid, e-3-2,4,5-trifluorophenyl prop-2-enoic acid, e-3-2,4,5-tris fluoranyl phenyl prop-2-enoic acid
5708777
(E)-3-(2,4,5-trifluorophenyl)prop-2-enoic acid
C1=C(C(=CC(=C1F)F)F)C=CC(=O)O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 184°C to 188°C
Quantity 1 g
Formula Weight 202.13
Percent Purity ≥97%
Chemical Name or Material 2,4,5-Trifluorocinnamic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'