Learn More
2,4,5-Trichlorophenol, TRC
Click to view available options
Quantity:
1 g
Unit Size:
1g
Specifications
Specifications
| Chemical Name or Material | 2,4,5-Trichlorophenol |
| CAS | 95-95-4 |
| Molecular Formula | C6 H3 Cl3 O |
| Quantity | 1 g |
| Synonym | Phenol','2,4,5-trichloro-, 2,4,5-Trichlorophenol, 2.4,5-Trichlorophenic acid, Dowicide 2, NSC 2266, Preventol I, TCP |
| InChI Formula | InChI=1S/C6H3Cl3O/c7-3-1-5(9)6(10)2-4(3)8/h1-2,10H |
| SMILES | Oc1cc(Cl)c(Cl)cc1Cl |
| IUPAC Name | 2,4,5-trichlorophenol |
| Molecular Weight (g/mol) | 197.45 |
| Formula Weight | 195.9249 |