Learn More
2,4,5-Trichloroaniline, 97%
Product Code. 11414587
Change view
Click to view available options
Quantity:
50 g
250 g
Unit Size:
250g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11414587 250 g 250g
11404587 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11414587 Supplier Thermo Scientific Alfa Aesar Supplier No. B21113.30
Request Bulk or Custom Format
CAS: 636-30-6 | C6H4Cl3N | 196.455 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 636-30-6
Molecular Formula C6H4Cl3N
Molecular Weight (g/mol) 196.455
MDL Number MFCD00007662
InChI Key GUMCAKKKNKYFEB-UHFFFAOYSA-N
Synonym benzenamine, 2,4,5-trichloro, unii-7xkk8s8wo0, aniline, 2,4,5-trichloro, 7xkk8s8wo0, 2,4,5-trichlorophenylamine, ccris 2882, pubchem3213, 2,5-trichloroaniline, aniline,4,5-trichloro, 2,4,5-trichloro aniline
PubChem CID 12487
IUPAC Name 2,4,5-trichloroaniline
SMILES C1=C(C(=CC(=C1Cl)Cl)Cl)N
Specifications
Melting Point 92°C to 95°C
Quantity 250 g
Odor Characteristic
UN Number UN2018
Beilstein 879091
Boiling Point 269°C to 270°C
Formula Weight 196.46
Percent Purity 97%
Chemical Name or Material 2,4,5-Trichloroaniline