Learn More
2-(3-Methoxyphenoxy)ethanamine, TRC
Specifications
Specifications
| Chemical Name or Material | 2-(3-Methoxyphenoxy)ethanamine |
| CAS | 6487-86-1 |
| Molecular Formula | C9H13NO2 |
| Quantity | 100 mg |
| Synonym | 2-(m-Methoxyphenoxy)ethylamine |
| InChI Formula | InChI=1S/C9H13NO2/c1-11-8-3-2-4-9(7-8)12-6-5-10/h2-4,7H,5-6,10H2,1H3 |
| SMILES | COc1cccc(OCCN)c1 |
| IUPAC Name | 2-(3-methoxyphenoxy)ethanamine |
| Molecular Weight (g/mol) | 167.21 |
| Formula Weight | 167.0946 |