Learn More
2,3-Hexanedione, 94%
Product Code. p-7044113
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
Product Code. 11341657
Brand: Thermo Scientific Alfa Aesar L04669.06
Request Bulk or Custom Format
CAS: 3848-24-6 | C6H10O2 | 114.14 g/mol
2,3-Hexanedione is used to evaluate the influence of xanthan concentration on the release of aroma compounds in xanthan-thickened food model systems. It reacts with ethylenediamine to yield macrocyclic tetradentate 12-membered nitrogen donor (N4) ligand.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2,3-Hexanedione is used to evaluate the influence of xanthan concentration on the release of aroma compounds in xanthan-thickened food model systems. It reacts with ethylenediamine to yield macrocyclic tetradentate 12-membered nitrogen donor (N4) ligand.
Solubility
Partly miscible in water.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 3848-24-6
Molecular Formula C6H10O2
Molecular Weight (g/mol) 114.14
MDL Number MFCD00009398
InChI Key MWVFCEVNXHTDNF-UHFFFAOYSA-N
Synonym 2,3-hexanedione, acetylbutyryl, acetyl butyryl, methyl propyl diketone, butyryl acetyl, hexanedione, unii-559anr3nvs, 2,3-hexandione, fema no. 2558, 559anr3nvs
PubChem CID 19707
ChEBI CHEBI:87583
IUPAC Name hexane-2,3-dione
SMILES CCCC(=O)C(C)=O
Specifications
Melting Point -30°C
Density 0.937
Boiling Point 127°C to 128°C
Flash Point 28°C (82°F)
Odor Characteristic
Refractive Index 1.412
Quantity 5 g
UN Number UN1224
Beilstein 1699896
Solubility Information Partly miscible in water.
Formula Weight 114.15
Percent Purity 94%
Chemical Name or Material 2,3-Hexanedione
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2,3-Hexanedione
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : 2,3-Hexanedione