Learn More
2,3-Epoxypropyl Phenyl Ether, Sigma-Aldrich™
Specifications
Specifications
| CAS | 122-60-1 |
| Melting Point | 3.5°C |
| Density | 1.11 g/cm3 at 20°C |
| MDL Number | MFCD00005133 |
| Quantity | 250 mL |
| Synonym | 2,3-Epoxypropyl phenyl ether, Glycidyl phenyl ether, 1,2-Epoxy-3-phenoxypropane, Phenyl 2,3-epoxypropyl ether, Phenyl glycid ether, 2,3-Epoxypropyl phenyl ether |
| Molecular Weight (g/mol) | 150.17 |
| Grade | Research |