Learn More
2,3-Diphenylquinoxaline, 98+%, Thermo Scientific Chemicals
Catalog Number 11460611
Click to view available options
Quantity:
5 g
25 g
100 g
1684-14-6
C20H14N2
282.346
MFCD00014659
RSNQVABHABAKEZ-UHFFFAOYSA-N
74311
2,3-diphenylquinoxaline
C1=CC=C(C=C1)C2=NC3=CC=CC=C3N=C2C4=CC=CC=C4
1684-14-6
C20H14N2
282.346
MFCD00014659
RSNQVABHABAKEZ-UHFFFAOYSA-N
74311
2,3-diphenylquinoxaline
C1=CC=C(C=C1)C2=NC3=CC=CC=C3N=C2C4=CC=CC=C4
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Melting Point 124°C to 128°C
Quantity 5 g
Beilstein 209272
Synonym quinoxaline, 2,3-diphenyl, quinoxaline,2,3-diphenyl, 2,3-diphenyl-quinoxaline, diphenylquinoxaline, quinoxaline,3-diphenyl, acmc-1c07k, 2,3-diphenylguinoxaline, 2,3-diphenyl-1,4-benzodiazine
Formula Weight 282.35
Percent Purity ≥98%
Chemical Name or Material 2,3-Diphenylquinoxaline