missing translation for 'onlineSavingsMsg'
Learn More

2,3-Diphenyl-1-indenone, 97%, Thermo Scientific™

Product Code. 10615732 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
Packaging:
Glass bottle
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10615732 10 g Glass bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10615732 Supplier Thermo Scientific Acros Supplier No. 117320100
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 1801-42-9
CAS Min % 96.0
CAS Max % 100.0
Melting Point 151°C to 155°C
Color Red
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Molecular Formula C21H14O
MDL Number MFCD00003784
Quantity 10 g
Beilstein 07, II, 501
Synonym 2,3-diphenyl-1-indenone, 2,3-diphenyl-1h-inden-1-one, 2,3-diphenylindone, 2,3-diphenyl-1-indanone, unii-h3l707j8y8, 2,3-diphenyl-inden-1-one, indone,3-diphenyl, 1h-inden-1-one,3-diphenyl, indone, 2,3-diphenyl, 2,3-diphenylindenone
InChI Key POVRXYLNLVVNGW-UHFFFAOYSA-N
SMILES C1=CC=C(C=C1)C2=C(C(=O)C3=CC=CC=C32)C4=CC=CC=C4
IUPAC Name 2,3-diphenylinden-1-one
Molecular Weight (g/mol) 282.33
PubChem CID 74536
ChEBI CHEBI:34231
Formula Weight 282.33
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 2, 3-Diphenyl-1-indenone, 97%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.