missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dimethylhexane, Thermo Scientific™

Product Code. 17565585 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 mL
5 mL
Unit Size:
1mL
5mL
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

Product Code. 17565585

Brand: Thermo Scientific Acros 469270050

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

Chemical Identifiers

CAS 584-94-1
Molecular Formula C8H18
Molecular Weight (g/mol) 114.23
MDL Number 00039951
InChI Key JXPOLSKBTUYKJB-UHFFFAOYNA-N
IUPAC Name 2,3-dimethylhexane
SMILES CCCC(C)C(C)C

Specifications

Alpha Vector DIMETHYLHEXANE
Color Clear colorless to almost colorless
Flash Point 7°C
Infrared Spectrum Conforms
Packaging Glass Bottle
Refractive Index 1.3990 to 1.4010 (20°C, 589 nm)
Quantity 5 mL
Physical Form Liquid
Chemical Name or Material 2,3-Dimethylhexane
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.