Learn More
2,3-Dimethylbutane, 99%
Product Code. p-7026293
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
Product Code. 11485856
Brand: Thermo Scientific Alfa Aesar B22223.06
Request Bulk or Custom Format
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
CAS: 79-29-8 | C6H14 | 86.178 g/mol
2,3-Dimethylbutane is a branched saturated hydrocarbon for proteomics research. It is used in organic synthesis, catalytic agent, petrochemical additive.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2,3-Dimethylbutane is a branched saturated hydrocarbon for proteomics research. It is used in organic synthesis, catalytic agent, petrochemical additive.
Solubility
Soluble in water (1mg/ml @22°C), DMSO, ethanol, acetone.
Notes
Incompatible with strong oxidizing agents, oxygen, ammonia.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 79-29-8
Molecular Formula C6H14
Molecular Weight (g/mol) 86.178
MDL Number MFCD00008925
InChI Key ZFFMLCVRJBZUDZ-UHFFFAOYSA-N
Synonym diisopropyl, biisopropyl, butane, 2,3-dimethyl, 1,1,2,2-tetramethylethane, hsdb 76, unii-68isq7a432, ccris 6020, ch3 2chch ch3 2, acmc-1bmfq, dsstox_cid_5112
PubChem CID 6589
IUPAC Name 2,3-dimethylbutane
SMILES CC(C)C(C)C
Specifications
Melting Point -129°C
Boiling Point 57°C to 59°C
Flash Point −29°C (−20°F)
Odor Mild
Quantity 5 g
UN Number UN2457
Beilstein 1730737
Formula Weight 86.18
Percent Purity 99%
Chemical Name or Material 2,3-Dimethylbutane