Learn More
2,3-Dimethylbutane, 98+%
Product Code. 15471867
Change view
Click to view available options
Quantity:
250 mL
Unit Size:
250mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15471867 250 mL 250mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Regulatory: This product is subject to excise duty on mineral oils in accordance with European Directive 2003/96/EC.
We regret to inform you that this product is not available for the Canary Islands, Ceuta and Melilla. We apologise for the inconvenience.
Product Code. 15471867 Supplier Thermo Scientific Alfa Aesar Supplier No. 043142.AK
 Restricted Item
Estimated Shipment: 14-12-2026
Add to basket
Regulatory: This product is subject to excise duty on mineral oils in accordance with European Directive 2003/96/EC.
We regret to inform you that this product is not available for the Canary Islands, Ceuta and Melilla. We apologise for the inconvenience.
CAS: 79-29-8 | C6H14 | 86.178 g/mol
2,3-Dimethylbutane is used in high octane fuels and in organic synthesis. It is also used as a gas chromatography standard.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 79-29-8
Molecular Formula C6H14
Molecular Weight (g/mol) 86.178
MDL Number MFCD00008925
InChI Key ZFFMLCVRJBZUDZ-UHFFFAOYSA-N
Synonym diisopropyl, biisopropyl, butane, 2,3-dimethyl, 1,1,2,2-tetramethylethane, hsdb 76, unii-68isq7a432, ccris 6020, ch3 2chch ch3 2, acmc-1bmfq, dsstox_cid_5112
PubChem CID 6589
IUPAC Name 2,3-dimethylbutane
SMILES CC(C)C(C)C
Specifications
Melting Point °C to 129°C
Density 0.662
Boiling Point 57°C to 59°C
Flash Point -29°C (-20°F)
Assay Percent Range >98%
Refractive Index 1.375
Quantity 250 mL
UN Number UN2457
Beilstein 1730737
Solubility Information Miscible with ethanol,ether and acetone. Immiscible with water.
Formula Weight 86.18
Percent Purity ≥98%
Physical Form Liquid
Chemical Name or Material 2, 3-Dimethylbutane