Learn More
2,3-Dimethyl-6-amino-2H-indazole Hydrochloride, Mikromol™
Product Code. 30464003
Click to view available options
Quantity:
25 mg
100 mg
4 x 25 mg
Unit Size:
100mg
25mg
Product Code. 30464003
Brand: Mikromol MM3391.0100254AMP
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
2,3-Dimethyl-6-amino-2H-indazole Hydrochloride, Mikromol
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 2,3-Dimethyl-6-amino-2H-indazole Hydrochloride
CAS 635702-60-2
Analyte or Component Names 2,3-Dimethyl-6-amino-2H-indazole Hydrochloride
Shipping Condition Room Temperature
Molecular Formula C9 H11 N3 . Cl H
Certifications/Compliance ISO 17025
Synonym 2H-Indazol-6-amine, 2,3-dimethyl-, hydrochloride (1:1), 2H-Indazol-6-amine, 2,3-dimethyl-, monohydrochloride (9CI), 2,3-Dimethyl-6-amino-2H-indazole hydrochloride
InChI Formula InChI=1S/C9H11N3.ClH/c1-6-8-4-3-7(10)5-9(8)11-12(6)2;/h3-5H,10H2,1-2H3;1H
Solution Type Neat
SMILES Cl.Cc1c2ccc(N)cc2nn1C
IUPAC Name 2,3-dimethylindazol-6-amine;hydrochloride
Molecular Weight (g/mol) 197.66
Quantity 4 x 25 mg
Formula Weight 197.072 g/mol