Learn More
2,3-Dimethyl-6-amino-2H-indazole Hydrochloride, Mikromol™
Click to view available options
Quantity:
25 mg
100 mg
4 x 25 mg
Unit Size:
100mg
25mg
Description
2,3-Dimethyl-6-amino-2H-indazole Hydrochloride
Specifications
Specifications
| Chemical Name or Material | 2,3-Dimethyl-6-amino-2H-indazole Hydrochloride |
| CAS | 635702-60-2 |
| Analyte or Component Names | 2,3-Dimethyl-6-amino-2H-indazole Hydrochloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C9 H11 N3 . Cl H |
| Certifications/Compliance | ISO 17025 |
| Synonym | 2H-Indazol-6-amine, 2,3-dimethyl-, hydrochloride (1:1), 2H-Indazol-6-amine, 2,3-dimethyl-, monohydrochloride (9CI), 2,3-Dimethyl-6-amino-2H-indazole hydrochloride |
| InChI Formula | InChI=1S/C9H11N3.ClH/c1-6-8-4-3-7(10)5-9(8)11-12(6)2;/h3-5H,10H2,1-2H3;1H |
| SMILES | Cl.Cc1c2ccc(N)cc2nn1C |
| IUPAC Name | 2,3-dimethylindazol-6-amine;hydrochloride |
| Show More |