missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2,3-Dihydroxypyridine, 98%

Product Code. 11387136
Change view
Click to view available options
Quantity:
25g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11387136 25g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11387136 Supplier Alfa Aesar™ Supplier No. L07104.14
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Applications
2,3-Dihydroxypyridine(DHP) is used in the synthesis of DHP loaded amberlite XAD-2 via azo coupler. It was used in the synthesis of new macromolecular chelators by loading DHP on cellulose via linkers -NH-CH2-CH2-NH-SO2-C6H4-N=N- and -SO2-C6H4-N=N.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible materials are Strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Specifications

CAS 16867-04-2
Molecular Formula C5H5NO2
MDL Number MFCD00006271
Quantity 25g
Beilstein 109848
Synonym 2,3-dihydroxypyridine, 2,3-pyridinediol, pyridine-2,3-diol, 3-hydroxy-2-pyridone, 2 1h-pyridinone, 3-hydroxy, 3-hydroxy-2 1h-pyridinone, 3-hydroxypyridin-2 1h-one, 2 1h-pyridone, 3-hydroxy, hydroxypyridone, unii-7hy4bcj130
InChI Key GGOZGYRTNQBSSA-UHFFFAOYSA-N
SMILES C1=CNC(=O)C(=C1)O
IUPAC Name 3-hydroxy-1H-pyridin-2-one
Molecular Weight (g/mol) 111.1
PubChem CID 28115
Formula Weight 111.1
Percent Purity 98%
Chemical Name or Material 2,3-Dihydroxypyridine
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.