missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dihydro-1-benzofuran-5-yl Isocyanate, TRC

Product Code. 30391367
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
10mg
Product Code. Quantity unitSize
30391367 100 mg -
30390848 50 mg -
30394569 10 mg 10mg
3 options
This item is not returnable. View return policy

Product Code. 30391367

Brand: Toronto Research Chemicals TRCI779418100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 215162-92-8
Formula Weight 161.0477
InChI Formula InChI=1S/C9H7NO2/c11-6-10-8-1-2-9-7(5-8)3-4-12-9/h1-2,5H,3-4H2
IUPAC Name 5-isocyanato-2,3-dihydro-1-benzofuran
Molecular Formula C9H7NO2
Molecular Weight (g/mol) 161.16
SMILES O=C=Nc1ccc2OCCc2c1
Chemical Name or Material 2,3-Dihydro-1-benzofuran-5-yl Isocyanate

Specifications

Quantity 100 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.