Learn More
2,3-Difluorophenylacetic acid, 98%
Product Code. 11412738
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
Product Code. 11412738
Brand: Thermo Scientific Alfa Aesar A16728.03
Request Bulk or Custom Format
CAS: 145689-41-4 | C8H6F2O2 | 172.131 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 145689-41-4
Molecular Formula C8H6F2O2
Molecular Weight (g/mol) 172.131
MDL Number MFCD00040968
InChI Key UXSQXUSJGPVOKT-UHFFFAOYSA-N
Synonym 2,3-difluorophenylacetic acid, 2-2,3-difluorophenyl acetic acid, 2,3-difluorophenyl acetic acid, 2,3-difluorophenylaceticacid, benzeneacetic acid, 2,3-difluoro, 2,3-difluorobenzeneacetic acid, rarechem al bo 0630, pubchem1916, acmc-1crot
PubChem CID 520772
IUPAC Name 2-(2,3-difluorophenyl)acetic acid
SMILES C1=CC(=C(C(=C1)F)F)CC(=O)O
Specifications
Melting Point 117°C to 119°C
Quantity 1 g
Formula Weight 172.13
Percent Purity 98%
Chemical Name or Material 2,3-Difluorophenylacetic acid
compliance-icons
Product Identifier
  • 2,3-Difluorophenylacetic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : 2,3-Difluorophenylacetic acid