Learn More
2,3-Dichloropyridine-4-boronic acid pinacol ester, 95%, Thermo Scientific™
Product Code. 15447695
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
Product Code. 15447695
Brand: Thermo Scientific Alfa Aesar H27498.MD
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1073353-78-2
Molecular Formula C11H14BCl2NO2
Molecular Weight (g/mol) 273.95
MDL Number MFCD06798257
InChI Key LRQNNFMOVWWIHA-UHFFFAOYSA-N
Synonym 2,3-dichloro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 2,3-dichloropyridine-4-boronic acid pinacol ester, pyridine, 2,3-dichloro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, abbypharma ap-13-5531, 2,3-dichloropyridine-4-boronic acid, pinacol ester, 2,3-dichloropyridin-4-yl boronic acid pinacol ester, 2,3-dichloro-4-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine
PubChem CID 44755158
IUPAC Name 2,3-dichloro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
SMILES CC1(C)OB(OC1(C)C)C1=C(Cl)C(Cl)=NC=C1
Specifications
Melting Point 83°C to 85°C
Quantity 250 mg
Formula Weight 273.96
Percent Purity 95%
Chemical Name or Material 2,3-Dichloropyridine-4-boronic acid pinacol ester