Learn More
2',3'-Dichloroacetophenone, 98%, Thermo Scientific Chemicals
Catalog Number 11420543
Click to view available options
Quantity:
1 g
5 g
25 g
56041-57-7
C8H6Cl2O
189.04
MFCD00052988
KMABBMYSEVZARZ-UHFFFAOYSA-N
1-2,3-dichlorophenyl ethanone, 2',3'-dichloroacetophenone, 2,3-dichloroacetophenone, 1-2,3-dichlorophenyl ethan-1-one, ethanone, 1-2,3-dichlorophenyl, 1-acetyl-2,3-dichlorobenzene, pubchem3368, 1-2,3-dichlorophenyl-1-ethanone, 1-2,3-bis chloranyl phenyl ethanone
91986
1-(2,3-dichlorophenyl)ethanone
CC(=O)C1=CC=CC(Cl)=C1Cl
56041-57-7
C8H6Cl2O
189.04
MFCD00052988
KMABBMYSEVZARZ-UHFFFAOYSA-N
1-2,3-dichlorophenyl ethanone, 2',3'-dichloroacetophenone, 2,3-dichloroacetophenone, 1-2,3-dichlorophenyl ethan-1-one, ethanone, 1-2,3-dichlorophenyl, 1-acetyl-2,3-dichlorobenzene, pubchem3368, 1-2,3-dichlorophenyl-1-ethanone, 1-2,3-bis chloranyl phenyl ethanone
91986
1-(2,3-dichlorophenyl)ethanone
CC(=O)C1=CC=CC(Cl)=C1Cl
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Density 1.293
Boiling Point 106°C to 108°C (2 mmHg)
Refractive Index 1.56
Quantity 1 g
Beilstein 2438815
Formula Weight 189.04
Percent Purity 98%
Chemical Name or Material 2',3'-Dichloroacetophenone