missing translation for 'onlineSavingsMsg'
Learn More

2,3-Bis(bromomethyl)quinoxaline, 97%, Thermo Scientific™

Product Code. 15424888
Click to view available options
Quantity:
2 g
10 g
Unit Size:
10g
2g
This item is not returnable. View return policy

Product Code. 15424888

Brand: Thermo Scientific Alfa Aesar H58649.09

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3138-86-1
Molecular Formula C10H8Br2N2
Molecular Weight (g/mol) 315.996
MDL Number MFCD00006729
InChI Key LHKFFORGJVELPC-UHFFFAOYSA-N
Synonym 2,3-bis bromomethyl quinoxaline, quinoxaline, 2,3-bis bromomethyl, 2,3-bis bromomethyl-1,4-benzodiazine, benzoic acid,3-fluoro-5-nitro, 2,4-benzodiazine, 2-hydroxybenzthiazole, acmc-1cq4g, 5-23-07-00271 beilstein handbook reference, ksc522c3j, quinoxaline,3-bis bromomethyl
PubChem CID 18449
IUPAC Name 2,3-bis(bromomethyl)quinoxaline
SMILES C1=CC=C2C(=C1)N=C(C(=N2)CBr)CBr

Specifications

Melting Point 152°C to 156°C
Quantity 10 g
UN Number UN3261
Formula Weight 315.99
Percent Purity 97%
Chemical Name or Material 2,3-Bis(bromomethyl)quinoxaline
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.