Learn More
2,3-Benzanthracene, 98%
Product Code. p-4509357 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
500 mg
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
500mg
5g
Product Code. Quantity Packaging unitSize
10293980 500 mg Glass bottle 500mg
10468050 1 g Glass bottle 1g
11954451 5 g Glass bottle 5g
3 options
Product Code. 10293980
Brand: Thermo Scientific Acros 105265000
Request Bulk or Custom Format
CAS: 92-24-0 | C18H12 | 228.29 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 92-24-0
Molecular Formula C18H12
Molecular Weight (g/mol) 228.29
InChI Key IFLREYGFSNHWGE-UHFFFAOYSA-N
Synonym naphthacene, 2,3-benzanthracene, rubene, benz b anthracene, 2,3-benzanthrene, chrysogen, hydrocarbon, unii-qyj5z6712r, ccris 1183
PubChem CID 7080
ChEBI CHEBI:32600
IUPAC Name tetracene
SMILES C1=CC=C2C=C3C=C4C=CC=CC4=CC3=CC2=C1
Specifications
Melting Point >300°C
Color Orange
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 500 mg
Beilstein 05,IV,2545
Merck Index 15,6454
Solubility Information Solubility in water: insoluble. Other solubilities: difficulty soluble in most solvents,soluble in hot xylene
Formula Weight 228.29
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 2, 3-Benzanthracene
Show More Show Less
compliance-icons
Product Identifier
  • 2,3-Benzanthracene
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
Hazard Statement
  • H410-Very toxic to aquatic life with long lasting effects.
  • H400-Very toxic to aquatic life.
Precautionary Statement