Learn More
2,3,4-Trifluorobenzoyl Chloride, TRC
Specifications
Specifications
| Chemical Name or Material | 2,3,4-Trifluorobenzoyl Chloride |
| CAS | 157373-08-5 |
| Molecular Formula | C7H2ClF3O |
| Quantity | 1 g |
| InChI Formula | InChI=1S/C7H2ClF3O/c8-7(12)3-1-2-4(9)6(11)5(3)10/h1-2H |
| SMILES | Fc1ccc(C(=O)Cl)c(F)c1F |
| IUPAC Name | 2,3,4-trifluorobenzoyl chloride |
| Molecular Weight (g/mol) | 194.54 |
| Formula Weight | 193.9746 |