Learn More
2,3,4-Trifluorobenzoyl chloride, 97%, Thermo Scientific™
Product Code. 11425487
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11425487 5 g 5g
11415487 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11425487 Supplier Thermo Scientific Alfa Aesar Supplier No. B23484.06
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 157373-08-5
Molecular Formula C7H2ClF3O
Molecular Weight (g/mol) 194.537
MDL Number MFCD00075078
InChI Key NXRQXCFBZGIRGN-UHFFFAOYSA-N
Synonym 2,3,4-trifluorobenzoylchloride, 2,3,4-trifluoro-benzoyl chloride, benzoyl chloride, 2,3,4-trifluoro, benzoyl chloride, 2,3,4-trifluoro-9ci, trifluorobenzoylchloride, trifluorobenzoyl chloride, intermediates-zcf02190, acmc-209dg2, 2,3,4-trifluorobenzoyl chloride
PubChem CID 2734022
IUPAC Name 2,3,4-trifluorobenzoyl chloride
SMILES C1=CC(=C(C(=C1C(=O)Cl)F)F)F
Specifications
Density 1.522
Boiling Point 143°C to 144°C (130 mmHg)
Flash Point 92°C (197°F)
Refractive Index 1.497
Quantity 5 g
UN Number UN3265
Beilstein 9122972
Sensitivity Moisture sensitive
Formula Weight 194.54
Percent Purity 97%
Chemical Name or Material 2,3,4-Trifluorobenzoyl chloride