missing translation for 'onlineSavingsMsg'
Learn More

2,3,4-Tri-O-benzyl-beta-L-arabinopyranose, 97%, Thermo Scientific™

Product Code. 11399745
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11399745 1 g 1g
11309755 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11399745 Supplier Thermo Scientific Alfa Aesar Supplier No. L19790.03
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
2,3,4-Tri-O-benzyl-beta-L-arabinopyranose is used as a pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 77943-33-0
Molecular Formula C26H28O5
Molecular Weight (g/mol) 420.505
MDL Number MFCD05664044
InChI Key HTSKDJMXBBFKKG-SSUZURRFSA-N
Synonym 2,3,4-tri-o-benzyl-beta-l-arabinopyranose, 2,3,4-tri-o-benzyl-b-l-arabinopyranose, 2s,3r,4s,5s-3,4,5-tris benzyloxy oxan-2-ol, 2s,3r,4s,5s-3,4,5-tris benzyloxy tetrahydro-2h-pyran-2-ol
PubChem CID 74892353
IUPAC Name (2S,3R,4S,5S)-3,4,5-tris(phenylmethoxy)oxan-2-ol
SMILES C1C(C(C(C(O1)O)OCC2=CC=CC=C2)OCC3=CC=CC=C3)OCC4=CC=CC=C4

Specifications

Melting Point 76°C to 88°C
Quantity 1 g
Solubility Information Insoluble in water.
Optical Rotation +78° (c=1 in Methanol)
Formula Weight 420.51
Percent Purity 97%
Chemical Name or Material 2,3,4-Tri-O-benzyl-beta-L-arabinopyranose
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.