missing translation for 'onlineSavingsMsg'
Learn More

2,3,4,5-Tetrafluorobenzotrifluoride, 97%, Thermo Scientific™

Catalog Number 11420267
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
654-53-5
C7HF7
218.074
MFCD01862013
ZWVOHERGWLDGFT-UHFFFAOYSA-N
2,3,4,5-tetrafluorobenzotrifluoride, 1,2,3,4-tetrafluoro-5-trifluoromethyl benzene, benzene, tetrafluoro trifluoromethyl, alpha,alpha,alpha,2,3,4,5-heptafluorotoluene, acmc-209trj, 3,4,5,6-tetrafluorobenzotrifluoride, 2,3,4,5-tetrafluoro-benzotrifluoride, 1,2,3,4-tetrafluoro-5-trifluoromethyl-benzene, 2,3,4,5-tetrafluoro-1-trifluoromethyl benzene
2733690
1,2,3,4-tetrafluoro-5-(trifluoromethyl)benzene
C1=C(C(=C(C(=C1F)F)F)F)C(F)(F)F
This item is not returnable. View return policy
654-53-5
C7HF7
218.074
MFCD01862013
ZWVOHERGWLDGFT-UHFFFAOYSA-N
2,3,4,5-tetrafluorobenzotrifluoride, 1,2,3,4-tetrafluoro-5-trifluoromethyl benzene, benzene, tetrafluoro trifluoromethyl, alpha,alpha,alpha,2,3,4,5-heptafluorotoluene, acmc-209trj, 3,4,5,6-tetrafluorobenzotrifluoride, 2,3,4,5-tetrafluoro-benzotrifluoride, 1,2,3,4-tetrafluoro-5-trifluoromethyl-benzene, 2,3,4,5-tetrafluoro-1-trifluoromethyl benzene
2733690
1,2,3,4-tetrafluoro-5-(trifluoromethyl)benzene
C1=C(C(=C(C(=C1F)F)F)F)C(F)(F)F

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Boiling Point 100°C to 102°C
Flash Point 22°C (71°F)
Refractive Index 1.3725
Quantity 5 g
UN Number UN1993
Beilstein 1973474
Formula Weight 218.07
Percent Purity 97%
Chemical Name or Material 2,3,4,5-Tetrafluorobenzotrifluoride
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'