missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2,3,4,5,6-Pentafluorophenoxyacetyl chloride, 99%

Catalog Number 11480517
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
55502-53-9
C8H2ClF5O2
260.544
MFCD01320685
WQLJMPAQIWVNGS-UHFFFAOYSA-N
pentafluorophenoxy acetyl chloride, 2,3,4,5,6-pentafluorophenoxyacetyl chloride, 2-perfluorophenoxy acetyl chloride, pentafluorophenoxyacetyl chloride, 2-pentafluorophenoxyacetyl chloride, 2-2,3,4,5,6-pentafluorophenoxy acetyl chloride, acmc-20aoj3, pentafluorophenyloxy-acetyl chloride, acetyl chloride, pentafluorophenoxy, 2,3,4,5,6-pentafluorophenoxyacetylchloride
97020
2-(2,3,4,5,6-pentafluorophenoxy)acetyl chloride
C(C(=O)Cl)OC1=C(C(=C(C(=C1F)F)F)F)F
This item is not returnable. View return policy
55502-53-9
C8H2ClF5O2
260.544
MFCD01320685
WQLJMPAQIWVNGS-UHFFFAOYSA-N
pentafluorophenoxy acetyl chloride, 2,3,4,5,6-pentafluorophenoxyacetyl chloride, 2-perfluorophenoxy acetyl chloride, pentafluorophenoxyacetyl chloride, 2-pentafluorophenoxyacetyl chloride, 2-2,3,4,5,6-pentafluorophenoxy acetyl chloride, acmc-20aoj3, pentafluorophenyloxy-acetyl chloride, acetyl chloride, pentafluorophenoxy, 2,3,4,5,6-pentafluorophenoxyacetylchloride
97020
2-(2,3,4,5,6-pentafluorophenoxy)acetyl chloride
C(C(=O)Cl)OC1=C(C(=C(C(=C1F)F)F)F)F
Boiling Point 116°C to 117°C (20 mmHg)
Quantity 1 g
UN Number UN3265
Sensitivity Moisture sensitive
Formula Weight 260.55
Percent Purity 99%
Chemical Name or Material 2,3,4,5,6-Pentafluorophenoxyacetyl chloride
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'