Learn More
(2,3,3a,4,7,7a-Hexahydro-2-methyl-1,3-dioxo-1H-isoindol-4-yl)carbamic Acid Phenylmethyl Ester-13C4, TRC
Specifications
Specifications
| Chemical Name or Material | (2,3,3a,4,7,7a-Hexahydro-2-methyl-1,3-dioxo-1H-isoindol-4-yl)carbamic Acid Phenylmethyl Ester-13C4 |
| Molecular Formula | C1313C4H18N2O4 |
| Quantity | 1 mg |
| InChI Formula | InChI=1S/C17H18N2O4/c1-19-15(20)12-8-5-9-13(14(12)16(19)21)18-17(22)23-10-11-6-3-2-4-7-11/h2-7,9,12-14H,8,10H2,1H3,(H,18,22)/i5+1,8+1,9+1,13+1 |
| SMILES | O=C1N(C)C(C2[13CH2][13CH]=[13CH][13CH](NC(OCC3=CC=CC=C3)=O)C21)=O |
| IUPAC Name | benzyl (2-methyl-1,3-dioxo-2,3,3a,4,7,7a-hexahydro-1H-isoindol-4-yl-4,5,6,7-13C4)carbamate |
| Molecular Weight (g/mol) | 318.31 |
| Formula Weight | 318.14 |