Learn More
2,2′-Thiodiacetic Acid, Sigma-Aldrich™
Specifications
Specifications
| CAS | 123-93-3 |
| Melting Point | 128°C to 131°C |
| Quantity | 250 g |
| Molecular Formula | S(CH2COOH)2 |
| MDL Number | MFCD00004359 |
| Synonym | 2,2′-Thiodiacetic acid, Thiodiglycollic acid, Bis(carboxymethyl)sulfide, 2,2′-Thiodiacetic acid |
| Molecular Weight (g/mol) | 150.15 |
| Chemical Name or Material | 2,2′-Thiodiacetic Acid |