Learn More
2-(2-Methyl-5-nitro-1H-imidazol-1-yl)acetic Acid, Mikromol™
Product Code. 17944026
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17944026 100 mg 100mg
17960596 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 17944026 Supplier Mikromol Supplier No. MM0012.09
 Restricted Item
Estimated Shipment: 03-08-2026
Add to basket
Add to basket
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
2-(2-Methyl-5-nitro-1H-imidazol-1-yl)acetic Acid
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 2-(2-Methyl-5-nitro-1H-imidazol-1-yl)acetic Acid
CAS 1010-93-1
Analyte or Component Names 2-(2-Methyl-5-nitro-1H-imidazol-1-yl)acetic Acid
Shipping Condition Room Temperature
Molecular Formula C6 H7 N3 O4
Certifications/Compliance ISO 17025
Synonym 2-(2-Methyl-5-nitro-1H-imidazol-1-yl)acetic Acid, Metronidazole Imp. G (EP), 2-Methyl-5-nitro-1H-imidazole-1-acetic acid, 1-De(2-hydroxyethyl)metronidazole-1-acetic acid, 2-Methyl-5-nitroimidazol-1-ylacetic acid, 2-Methyl-5-nitroimidazole-1-acetic acid, Metronidazoleacetic acid, Metronidazole Impurity G
InChI Formula InChI=1S/C6H7N3O4/c1-4-7-2-5(9(12)13)8(4)3-6(10)11/h2H,3H2,1H3,(H,10,11)
SMILES Cc1ncc([N+](=O)[O-])n1CC(=O)O
IUPAC Name 2-(2-methyl-5-nitroimidazol-1-yl)acetic acid
Molecular Weight (g/mol) 185.14
Quantity 100 mg
Formula Weight 185.0437 g/mol