missing translation for 'onlineSavingsMsg'
Learn More

2-(2-Ethylhexyloxy)-5-methoxy-1,4-benzenediacetonitrile, 98%, Thermo Scientific™

Catalog Number p-7033862
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
213749-91-8
C19H26N2O2
314.429
MFCD03701544
YGAQISIJKKIHRN-UHFFFAOYSA-N
2-2-ethylhexyloxy-5-methoxy-1,4-benzenediacetonitrile, 2,2'-2-2-ethylhexyloxy-5-methoxy-1,4-phenylene diacetonitrile, 2,2'-2-2-ethylhexyl oxy-5-methoxy-1,4-phenylene diacetonitrile, 2-4-cyanomethyl-5-2-ethylhexyl oxy-2-methoxyphenyl acetonitrile, 2-methoxy-5-2-ethylhexyloxy benzene-1, 2-methoxy-5-2'-ethylhexyloxy benzene-1, 1,4-bis cyanomethyl-2 2'-ethylhexyloxy-5-methoxybenzene, 2-4-cyanomethyl-2-2-ethylhexoxy-5-methoxyphenyl acetonitrile, 2-methoxy-5-2'-ethylhexyloxy benzene-1,4-diacetonitrile
4115353
2-[4-(cyanomethyl)-2-(2-ethylhexoxy)-5-methoxyphenyl]acetonitrile
CCCCC(CC)COC1=CC(=C(C=C1CC#N)OC)CC#N
This item is not returnable. View return policy
213749-91-8
C19H26N2O2
314.429
MFCD03701544
YGAQISIJKKIHRN-UHFFFAOYSA-N
2-2-ethylhexyloxy-5-methoxy-1,4-benzenediacetonitrile, 2,2'-2-2-ethylhexyloxy-5-methoxy-1,4-phenylene diacetonitrile, 2,2'-2-2-ethylhexyl oxy-5-methoxy-1,4-phenylene diacetonitrile, 2-4-cyanomethyl-5-2-ethylhexyl oxy-2-methoxyphenyl acetonitrile, 2-methoxy-5-2-ethylhexyloxy benzene-1, 2-methoxy-5-2'-ethylhexyloxy benzene-1, 1,4-bis cyanomethyl-2 2'-ethylhexyloxy-5-methoxybenzene, 2-4-cyanomethyl-2-2-ethylhexoxy-5-methoxyphenyl acetonitrile, 2-methoxy-5-2'-ethylhexyloxy benzene-1,4-diacetonitrile
4115353
2-[4-(cyanomethyl)-2-(2-ethylhexoxy)-5-methoxyphenyl]acetonitrile
CCCCC(CC)COC1=CC(=C(C=C1CC#N)OC)CC#N

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 1 g
UN Number UN3439
Formula Weight 314.42
Percent Purity 98%
Chemical Name or Material 2-(2-Ethylhexyloxy)-5-methoxy-1,4-benzenediacetonitrile
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'