Learn More
2,2-Dimethylbutanoyl Chloride, TRC
Product Code. 30384653
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30384653 5 g 5g
30372377 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30384653 Supplier Toronto Research Chemicals Supplier No. TRCD4714655G
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 2,2-Dimethylbutanoyl Chloride
CAS 5856-77-9
Molecular Formula C6H11ClO
Quantity 5 g
Synonym 2,2-Dimethylbutanoyl Chloride, 2,2-Dimethylbutyric Acid Chloride, 2,2-Dimethylbutyryl Chloride
InChI Formula InChI=1S/C6H11ClO/c1-4-6(2,3)5(7)8/h4H2,1-3H3
SMILES O=C(Cl)C(C)(C)CC
IUPAC Name 2,2-dimethylbutanoyl chloride
Molecular Weight (g/mol) 134.6
Formula Weight 134.05