missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-[2-(Dimethylamino)ethoxy]aniline, 97%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
Specifications
Specifications
| Assay Percent Range | 97% |
| Packaging | Amber Glass Bottle |
| Quantity | 5 g |
| Formula Weight | 180.25 |
| Percent Purity | 97% |
| Chemical Name or Material | 2-[2-(Dimethylamino)ethoxy]aniline |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction