Learn More
2,2-Dichloro-1,1-difluoroethyl methyl ether, 97%, Thermo Scientific Chemicals
Product Code. 11314026
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11314026 5 g 5g
11324026 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11314026 Supplier Thermo Scientific Alfa Aesar Supplier No. L17285.06
 Restricted Item
In Stock
Add to basket
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2,2-Dichloro-1,1-difluoroethyl methyl ether is used as a lipid soluble anesthetic agent. It is a neuromuscular blocking agent given concurrently to get the desired degree of muscular relaxation. Further, it is a powerful analgesic agent.
Solubility
Miscible with alcohol, acetone, chloroform, ether, fixed oils and benzene. Immiscible with water.
Notes
Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 76-38-0
Molecular Formula C3H4Cl2F2O
Molecular Weight (g/mol) 164.96
MDL Number MFCD00040144
InChI Key RFKMCNOHBTXSMU-UHFFFAOYSA-N
Synonym methoxyflurane, methoxyfluran, penthrane, methoflurane, metoxifluran, metoxfluran, methoxane, anecotan, metofane, pentrane
PubChem CID 4116
ChEBI CHEBI:6843
IUPAC Name 2,2-dichloro-1,1-difluoro-1-methoxyethane
SMILES COC(F)(F)C(Cl)Cl
Specifications
Melting Point -36°C
Density 1.44
Boiling Point 105°C
Flash Point 37°C (98°F)
Quantity 5 g
UN Number UN3271
Beilstein 1737766
Merck Index 14,5994
Solubility Information Miscible with alcohol,acetone,chloroform,ether,fixed oils and benzene. Immiscible with water.
Formula Weight 164.97
Percent Purity 97%
Chemical Name or Material 2,2-Dichloro-1,1-difluoroethyl methyl ether