missing translation for 'onlineSavingsMsg'
Learn More

2-(2-Chloroethoxy)ethanol, 99%

Product Code. 11414394
Click to view available options
Quantity:
50 g
250 g
1000 g
Unit Size:
1000g
250g
50g
This item is not returnable. View return policy

Product Code. 11414394

Brand: Thermo Scientific Alfa Aesar A13074.0B

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 628-89-7 | C4H9ClO2 | 124.56 g/mol

2-(2-Chloroethoxy)ethanol is employed in the preparation of o-nitrophenylbromoacetaldehyde bis-2-(2-chloroethoxy)-ethyl acetal, quetiapine and 2-(2-azidoethoxy)ethanol.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-(2-Chloroethoxy)ethanol is employed in the preparation of o-nitrophenylbromoacetaldehyde bis-2-(2-chloroethoxy)-ethyl acetal, quetiapine and 2-(2-azidoethoxy)ethanol.

Solubility
Miscible with water.

Notes
Moisture sensitive. Store in a cool place. Incompatible with strong oxidizing agents and strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 628-89-7
Molecular Formula C4H9ClO2
Molecular Weight (g/mol) 124.56
MDL Number MFCD00002870
InChI Key LECMBPWEOVZHKN-UHFFFAOYSA-N
Synonym 2-2-chloroethoxy ethanol, ethanol, 2-2-chloroethoxy, diglycol chlorohydrin, diglycol chlorhydrin, 2-chloroethyl 2-hydroxyethyl ether, monochloropolyoxyethylene, 2-2-chloroethoxy ethan-1-ol, unii-r2up818l4i, diethylene glycol monochlorohydrin, polyethylene glycol chloride 210
PubChem CID 12361
IUPAC Name 2-(2-chloroethoxy)ethanol
SMILES OCCOCCCl

Specifications

Density 1.18
Boiling Point 185°C
Flash Point 90°C (194°F)
Linear Formula ClCH2CH2OCH2CH2OH
Refractive Index 1.4535
Quantity 1000 g
Beilstein 506015
Solubility Information Miscible with water.
Formula Weight 124.57
Percent Purity 99%
Chemical Name or Material 2-(2-Chloroethoxy)ethanol
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.