Learn More
2,2-Bis(hydroxyethyl)-(iminotris)- (Hydroxymethyl)-methane, Millipore™
Product Code. 10625341
Change view
Click to view available options
Quantity:
250 g
Unit Size:
250g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10625341 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10625341 Supplier Millipore Supplier No. 1032520250
 Restricted Item
Estimated Shipment: 26-11-2026
BIS-TRIS
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 6976-37-0
Grade Research
Molecular Weight (g/mol) 209.24
Specifications
Quantity 250 g
Solution Type Buffer
MDL Number MFCD00002853
Synonym 2,2-Bis(hydroxyethyl)-(iminotris)- (hydroxymethyl)-methane, 2,2-Bis(hydroxyethyl)-(iminotris)- (hydroxymethyl)-methane
compliance-icons
Product Identifier
  • 2,2-Bis(hydroxyethyl)-(iminotris)- (hydroxymethyl)-methane BIS-TRIS
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P264-Wash skin thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: 2-[Bis(2-hydroxyethyl)amino]-2-(hydroxymethyl)propane-1,3-diol